accessibility menu, dialog, popup

UserName

CH3NH(S)H3N

CAS RN 26116-12-1

IUPAC Name: (2S)-2-(azaniumylmethyl)-1-ethylpyrrolidin-1-ium
Synonyms: (2S)-2-(ammoniomethyl)-1-ethylpyrrolidin-1-ium
Molecular Weight (g/mol): 130.23
Molecular Formula: C7H18N2
InChi Key: UNRBEYYLYRXYCG-ZETCQYMHSA-P
SMILES: CC[NH+]1CCC[C@H]1C[NH3+]
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)