missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Aminomethyl)-1,3-dioxolane 98.0+%, TCI America™
$281.48 - $920.84
Chemical Identifiers
| CAS | 4388-97-0 |
|---|---|
| Molecular Formula | C4H9NO2 |
| Molecular Weight (g/mol) | 103.121 |
| MDL Number | MFCD00142862 |
| InChI Key | QXXRLUXAOFVOTE-UHFFFAOYSA-N |
| PubChem CID | 10855394 |
| IUPAC Name | 1,3-dioxolan-2-ylmethanamine |
| SMILES | C1COC(O1)CN |
Chemical Identifiers
| 4388-97-0 | |
| 103.121 | |
| QXXRLUXAOFVOTE-UHFFFAOYSA-N | |
| 1,3-dioxolan-2-ylmethanamine |
| C4H9NO2 | |
| MFCD00142862 | |
| 10855394 | |
| C1COC(O1)CN |
Specifications
| 4388-97-0 | |
| C4H9NO2 | |
| 1 g | |
| QXXRLUXAOFVOTE-UHFFFAOYSA-N | |
| 1,3-dioxolan-2-ylmethanamine | |
| 10855394 | |
| ≥98.0% (GC,T) | |
| 2-(Aminomethyl)-1,3-dioxolane |
| Colorless | |
| MFCD00142862 | |
| 1993 | |
| C1COC(O1)CN | |
| 103.121 | |
| 103.12 | |
| Liquid |
Safety and Handling
TSCA : No