accessibility menu, dialog, popup

UserName

OOH2N

CAS RN 4388-97-0

IUPAC Name: 1-(1,3-dioxolan-2-yl)methanamine
Synonyms: 1-(1,3-dioxolan-2-yl)methanamine
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: QXXRLUXAOFVOTE-UHFFFAOYSA-N
SMILES: NCC1OCCO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results