missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)-1,3-dioxolane 98.0+%, TCI America™

Catalog No. A11795G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11795G 5 g
A11791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11795G Supplier TCI America Supplier No. A11795G
Only null left

Chemical Identifiers

CAS 4388-97-0
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00142862
InChI Key QXXRLUXAOFVOTE-UHFFFAOYSA-N
PubChem CID 10855394
IUPAC Name 1,3-dioxolan-2-ylmethanamine
SMILES C1COC(O1)CN

Specifications

Color Colorless
Quantity 5 g
UN Number 1993
Formula Weight 103.12
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Aminomethyl)-1,3-dioxolane