missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoisobutyric Acid 98.0+%, TCI America™

Supplier:  TCI America A0323500G

 View more versions of this product

Catalog No. A0323500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Aminoisobutyric Acid
335°C
C4H9NO2
500 g
FUOOLUPWFVMBKG-UHFFFAOYSA-N
2-amino-2-methylpropanoic acid
6119
103.12
Crystalline Powder
62-57-7
White
MFCD00008049
2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid
CC(C)(C(=O)O)N
103.121
CHEBI:27971
≥98.0% (T)

Chemical Identifiers

62-57-7
103.121
FUOOLUPWFVMBKG-UHFFFAOYSA-N
6119
2-amino-2-methylpropanoic acid
C4H9NO2
MFCD00008049
2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid
CHEBI:27971
CC(C)(C(=O)O)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1214&(9)-2028

RTECSNumber : AY7000000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions