accessibility menu, dialog, popup

UserName

CAS RN 62-57-7

IUPAC Name: 2-amino-2-methylpropanoic acid
Synonyms: α-aminoisobutyric acid α-methylalanine 2MA AIB Aib 2-methylalanine
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: FUOOLUPWFVMBKG-UHFFFAOYSA-N
SMILES: CC(C)(N)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (5)