missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoisobutyric Acid 98.0+%, TCI America™

Catalog No. A032325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A032325G 25 g
A0323100G 100 g
A0323500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A032325G Supplier TCI America Supplier No. A032325G

Chemical Identifiers

CAS 62-57-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00008049
InChI Key FUOOLUPWFVMBKG-UHFFFAOYSA-N
Synonym 2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid
PubChem CID 6119
ChEBI CHEBI:27971
IUPAC Name 2-amino-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)N

Specifications

Melting Point 335°C
Color White
Quantity 25 g
Formula Weight 103.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoisobutyric Acid
EINECSNumber (2)-1214&(9)-2028
RTECSNumber AY7000000
TSCA Yes