Learn More
2-Aminoisobutyric acid, 99%
CAS: 62-57-7 | C4H9NO2 | 103.121 g/mol
Supplier: Thermo Scientific Chemicals A1302130
Description
2-Aminoisobutyric acid is a non- protein amino acid present in some antibiotics. It acts as a strong helix inducer in peptides. It is non-metabolisable used in bioassays.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Aminoisobutyric acid is a non- protein amino acid present in some antibiotics. It acts as a strong helix inducer in peptides. It is non-metabolisable used in bioassays.
Solubility
Soluble in water. Insoluble in alcohol and ether.
Notes
Incompatible with strong oxidizing agents.
Specifications
| 2-Aminoisobutyric acid | |
| >300°C | |
| MFCD00008049 | |
| 506496 | |
| 2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid | |
| FUOOLUPWFVMBKG-UHFFFAOYSA-N | |
| 2-amino-2-methylpropanoic acid | |
| 6119 | |
| 103.12 |
| 62-57-7 | |
| C4H9NO2 | |
| 250 g | |
| 14,445 | |
| Soluble in water. Insoluble in alcohol and ether. | |
| CC(C)(C(=O)O)N | |
| 103.121 | |
| CHEBI:27971 | |
| 99% |
Chemical Identifiers
| 62-57-7 | |
| 103.121 | |
| FUOOLUPWFVMBKG-UHFFFAOYSA-N | |
| 6119 | |
| 2-amino-2-methylpropanoic acid |
| C4H9NO2 | |
| MFCD00008049 | |
| 2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid | |
| CHEBI:27971 | |
| CC(C)(C(=O)O)N |
Safety and Handling
EINECSNumber : 200-544-0
RTECSNumber : AY7000000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only