missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminoisobutyric acid, 98+%
CAS: 62-57-7 | C4H9NO2 | 103.12 g/mol
$92.67 - $92.67
Chemical Identifiers
| CAS | 62-57-7 |
|---|---|
| Molecular Formula | C4H9NO2 |
| Molecular Weight (g/mol) | 103.12 |
| MDL Number | MFCD00008049 |
| InChI Key | FUOOLUPWFVMBKG-UHFFFAOYSA-N |
| Synonym | 2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid |
| PubChem CID | 6119 |
| ChEBI | CHEBI:27971 |
| IUPAC Name | 2-amino-2-methylpropanoic acid |
| SMILES | CC(C)(C(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC225760250
|
Thermo Scientific Chemicals
225760250 |
25 g |
Each for $92.67
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 62-57-7 | |
| 103.12 | |
| FUOOLUPWFVMBKG-UHFFFAOYSA-N | |
| 6119 | |
| 2-amino-2-methylpropanoic acid |
| C4H9NO2 | |
| MFCD00008049 | |
| 2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid | |
| CHEBI:27971 | |
| CC(C)(C(=O)O)N |
Specifications
| 62-57-7 | |
| 100.0 | |
| White | |
| 98+% | |
| C4H9NO2 | |
| MFCD00008049 | |
| 04, 414 | |
| 2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid | |
| FUOOLUPWFVMBKG-UHFFFAOYSA-N | |
| 2-amino-2-methylpropanoic acid | |
| 6119 | |
| 103.12 | |
| Crystalline Powder |
| 99.0 | |
| 335.0°C | |
| Authentic | |
| Glass bottle | |
| (CH3)2C(NH2)CO2H | |
| 25 g | |
| 15, 442 | |
| Solubility in water: soluble. Other solubilities: difficultly soluble in alcohol, insoluble in ether | |
| CC(C)(C(=O)O)N | |
| 103.12 | |
| CHEBI:27971 | |
| 98+% | |
| 2-Aminoisobutyric acid, ≥98% |
Safety and Handling
EINECSNumber : 200-544-0
RTECSNumber : AY7000000
TSCA : TSCA
RUO – Research Use Only