missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoisobutyric acid, 98+%

Catalog No. AC225760250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC225760250 25 g
AC225761000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC225760250 Supplier Thermo Scientific Chemicals Supplier No. 225760250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 62-57-7 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62-57-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00008049
InChI Key FUOOLUPWFVMBKG-UHFFFAOYSA-N
Synonym 2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid
PubChem CID 6119
ChEBI CHEBI:27971
IUPAC Name 2-amino-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 335.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (CH3)2C(NH2)CO2H
Quantity 25 g
Beilstein 04, 414
Merck Index 15, 442
Solubility Information Solubility in water: soluble. Other solubilities: difficultly soluble in alcohol, insoluble in ether
Formula Weight 103.12
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoisobutyric acid, ≥98%
Show More Show Less
EINECSNumber 200-544-0
RTECSNumber AY7000000
TSCA TSCA

RUO – Research Use Only