missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoindane hydrochloride, 98%

Catalog No. AAL1408306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1408306 5 g
AAL1408303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1408306 Supplier Thermo Scientific Chemicals Supplier No. L1408306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2338-18-3 | C9H11ClN | 168.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2338-18-3
Molecular Formula C9H11ClN
Molecular Weight (g/mol) 168.64
MDL Number MFCD00012549
InChI Key DLYRFJDJKGGVKT-UHFFFAOYSA-N
Synonym 2-aminoindan hydrochloride, 2-aminoindane hydrochloride, 2,3-dihydro-1h-inden-2-amine hydrochloride, 2-aminoindane hcl, 2-indanamine hydrochloride, 2-indanamine, hydrochloride, 2-indanylamine hydrochloride, su 8629 hydrochloride, indan-2-ylammonium chloride, indan-2-ylamine hydrochloride
PubChem CID 122764
IUPAC Name 2,3-dihydro-1H-inden-2-amine;hydrochloride
SMILES [Cl].NC1CC2=CC=CC=C2C1

Specifications

Melting Point 243°C to 248°C
Odor Odorless
Quantity 5 g
Beilstein 3913700
Formula Weight 169.66
Percent Purity 98%
Chemical Name or Material 2-Aminoindane hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 219-048-0
RTECSNumber NK4050000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only