missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminoindan 99.0+%, TCI America™
$169.63 - $589.05
Chemical Identifiers
| CAS | 2975-41-9 |
|---|---|
| Molecular Formula | C9H12ClN |
| Molecular Weight (g/mol) | 169.65 |
| MDL Number | MFCD00082597 |
| InChI Key | XEHNLVMHWYPNEQ-UHFFFAOYSA-N |
| Synonym | 2-aminoindan, 2-aminoindane, indan-2-amine, 2-indanamine, 2,3-dihydro-1h-inden-2-ylamine, indan-2-ylamine, indane-2-ylamine, 2-indanylamine, unii-1p810sq3ey, 1h-inden-2-amine, 2,3-dihydro |
| PubChem CID | 76310 |
| IUPAC Name | hydrogen 2,3-dihydro-1H-inden-2-amine chloride |
| SMILES | [H+].[Cl-].NC1CC2=CC=CC=C2C1 |
Chemical Identifiers
| 2975-41-9 | |
| 169.65 | |
| XEHNLVMHWYPNEQ-UHFFFAOYSA-N | |
| 76310 | |
| [H+].[Cl-].NC1CC2=CC=CC=C2C1 |
| C9H12ClN | |
| MFCD00082597 | |
| 2-aminoindan, 2-aminoindane, indan-2-amine, 2-indanamine, 2,3-dihydro-1h-inden-2-ylamine, indan-2-ylamine, indane-2-ylamine, 2-indanylamine, unii-1p810sq3ey, 1h-inden-2-amine, 2,3-dihydro | |
| hydrogen 2,3-dihydro-1H-inden-2-amine chloride |
Specifications
| 2975-41-9 | |
| 229°C | |
| MFCD00082597 | |
| 2735 | |
| XEHNLVMHWYPNEQ-UHFFFAOYSA-N | |
| hydrogen 2,3-dihydro-1H-inden-2-amine chloride | |
| 76310 | |
| ≥99.0% (GC,T) | |
| 2-Aminoindan |
| Brown | |
| C9H12ClN | |
| 5 g | |
| 2-aminoindan, 2-aminoindane, indan-2-amine, 2-indanamine, 2,3-dihydro-1h-inden-2-ylamine, indan-2-ylamine, indane-2-ylamine, 2-indanylamine, unii-1p810sq3ey, 1h-inden-2-amine, 2,3-dihydro | |
| [H+].[Cl-].NC1CC2=CC=CC=C2C1 | |
| 169.65 | |
| 133.19 | |
| Liquid |
Safety and Handling
TSCA : No