missing translation for 'onlineSavingsMsg'
Learn More

2-Aminofluorene 98.0+%, TCI America™

Catalog No. p-7116969
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06215G 5 g
A062125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A06215G Supplier TCI America Supplier No. A06215G
Only null left

Chemical Identifiers

CAS 153-78-6
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.238
MDL Number MFCD00001125
InChI Key CFRFHWQYWJMEJN-UHFFFAOYSA-N
Synonym 2-aminofluorene, 2-fluorenamine, aminofluoren, 2-fluoreneamine, fluorene, 2-amino, fluoren-2-amine, 2-fluorenylamine, 2-fluroenylamine, fluoren-2-ylamine, aminofluoren german
PubChem CID 1539
IUPAC Name 9H-fluoren-2-amine
SMILES C1C2=CC=CC=C2C3=C1C=C(C=C3)N

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 5 g
Formula Weight 181.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminofluorene
RTECSNumber LL5075000
TSCA No