missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethyl Isopropyl Ether 98.0+%, TCI America™

Catalog No. A12965ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12965ML 5 mL
A129625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12965ML Supplier TCI America Supplier No. A12965ML

Chemical Identifiers

CAS 81731-43-3
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00191603
InChI Key USECIYVEPXUVHT-UHFFFAOYSA-N
Synonym 2-Isopropoxyethylamine
PubChem CID 533866
IUPAC Name 2-(propan-2-yloxy)ethan-1-amine
SMILES CC(C)OCCN

Specifications

Color Yellow
Boiling Point 120°C
Quantity 5 mL
UN Number 2733
Formula Weight 103.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Aminoethyl Isopropyl Ether