accessibility menu, dialog, popup

UserName

CAS RN 81731-43-3

H3CCH3ONH2

CAS RN 81731-43-3

IUPAC Name: 2-(propan-2-yloxy)ethan-1-amine
Synonyms: 2-isopropoxyethanamine
Molecular Weight (g/mol): 103.17
Molecular Formula: C5H13NO
InChi Key: USECIYVEPXUVHT-UHFFFAOYSA-N
SMILES: CC(C)OCCN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results