Learn More
2-Aminoethyl diphenylborinate, 98%
CAS: 524-95-8 | C14H16BNO | 225.10 g/mol
Supplier: Thermo Scientific Chemicals A1660606
Description
2-Aminoethyl diphenylborinate is used for the densitometric determination of flavonoids and TRP activator. It is also used to manipulate intracellular release of calcium ions and in preparation of apoptotic inducers. It serves as a reactant for preparation of apoptotic inducers and in membrane-permeable SOCE (store operated calcuym entry) inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Aminoethyl diphenylborinate is used for the densitometric determination of flavonoids and TRP activator. It is also used to manipulate intracellular release of calcium ions and in preparation of apoptotic inducers. It serves as a reactant for preparation of apoptotic inducers and in membrane-permeable SOCE (store operated calcuym entry) inhibitor.
Solubility
Soluble in ethanol and methanol.
Notes
Incompatible with strong oxidizing agents.
Specifications
| 2-Aminoethyl diphenylborinate | |
| 187°C to 190°C | |
| MFCD00014823 | |
| 2942273 | |
| Soluble in ethanol and methanol. | |
| NCCOB(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 225.10 | |
| 225.1 |
| 524-95-8 | |
| C14H16BNO | |
| 5 g | |
| 2-aminoethyl diphenylborinate, 2-apb, 2-aminoethoxy diphenylborane, 2-aminoethoxydiphenylborane, 2-aminoethoxydiphenylborate, 2-aminoethoxydiphenyl borate, o-2-aminoethyl diphenylborinic acid, b-2-aminoethoxy diphenylborane, 2-diphenylboryl oxy ethanamine, borinic acid, o-2-aminoethyl diphenyl | |
| BLZVCIGGICSWIG-UHFFFAOYSA-N | |
| 2-aminoethyl diphenylborinate | |
| 1598 | |
| 98% |
Chemical Identifiers
| 524-95-8 | |
| 225.10 | |
| BLZVCIGGICSWIG-UHFFFAOYSA-N | |
| 1598 |
| C14H16BNO | |
| MFCD00014823 | |
| 2-aminoethyl diphenylborinate, 2-apb, 2-aminoethoxy diphenylborane, 2-aminoethoxydiphenylborane, 2-aminoethoxydiphenylborate, 2-aminoethoxydiphenyl borate, o-2-aminoethyl diphenylborinic acid, b-2-aminoethoxy diphenylborane, 2-diphenylboryl oxy ethanamine, borinic acid, o-2-aminoethyl diphenyl | |
| NCCOB(C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-366-5
RTECSNumber : ED6150000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only