missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethyl diphenylborinate, 98%

Catalog No. p-7023249
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1660606 5 g
AAA1660614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1660606 Supplier Thermo Scientific Chemicals Supplier No. A1660606

CAS: 524-95-8 | C14H16BNO | 225.10 g/mol

2-Aminoethyl diphenylborinate is used for the densitometric determination of flavonoids and TRP activator. It is also used to manipulate intracellular release of calcium ions and in preparation of apoptotic inducers. It serves as a reactant for preparation of apoptotic inducers and in membrane-permeable SOCE (store operated calcuym entry) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminoethyl diphenylborinate is used for the densitometric determination of flavonoids and TRP activator. It is also used to manipulate intracellular release of calcium ions and in preparation of apoptotic inducers. It serves as a reactant for preparation of apoptotic inducers and in membrane-permeable SOCE (store operated calcuym entry) inhibitor.

Solubility
Soluble in ethanol and methanol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 524-95-8
Molecular Formula C14H16BNO
Molecular Weight (g/mol) 225.10
MDL Number MFCD00014823
InChI Key BLZVCIGGICSWIG-UHFFFAOYSA-N
Synonym 2-aminoethyl diphenylborinate, 2-apb, 2-aminoethoxy diphenylborane, 2-aminoethoxydiphenylborane, 2-aminoethoxydiphenylborate, 2-aminoethoxydiphenyl borate, o-2-aminoethyl diphenylborinic acid, b-2-aminoethoxy diphenylborane, 2-diphenylboryl oxy ethanamine, borinic acid, o-2-aminoethyl diphenyl
PubChem CID 1598
SMILES NCCOB(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 187°C to 190°C
Quantity 5 g
Beilstein 2942273
Solubility Information Soluble in ethanol and methanol.
IUPAC Name 2-aminoethyl diphenylborinate
Formula Weight 225.1
Percent Purity 98%
Chemical Name or Material 2-Aminoethyl diphenylborinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-366-5
RTECSNumber ED6150000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only