missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethanethiol Hydrochloride 95.0+%, TCI America™

Catalog No. p-7116812
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A029625G 25 g
A0296100G 100 g
A0296500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A029625G Supplier TCI America Supplier No. A029625G

Chemical Identifiers

CAS 156-57-0
Molecular Formula C2H8ClNS
Molecular Weight (g/mol) 113.60
MDL Number MFCD00012904
InChI Key OGMADIBCHLQMIP-UHFFFAOYSA-N
Synonym cysteamine hydrochloride, 2-aminoethanethiol hydrochloride, cysteamine hcl, cysteaminium chloride, 2-mercaptoethylamine hydrochloride, 2-mercaptoethylamine hcl, bekaptan, mercaptamine hydrochloride, merkamin hydrochloride
PubChem CID 9082
IUPAC Name hydrogen 2-aminoethane-1-thiol chloride
SMILES [H+].[Cl-].NCCS

Specifications

Melting Point 68°C
Color White
pH 3.5 to 4.5 (40w/v% H2O soln.)
Quantity 25 g
Formula Weight 113.60
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoethanethiol Hydrochloride
EINECSNumber (9)-1422
RTECSNumber KJ0200000
TSCA Yes
Recommended Storage Refrigerator