missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethanethiol hydrochloride, 98%

Catalog No. AC153775000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153775000 500 g Glass bottle
AC153770050 5 g Glass bottle
AC153770250 25 g Glass bottle
AC153771000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC153775000 Supplier Thermo Scientific Chemicals Supplier No. 153775000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 156-57-0 | C2H8ClNS | 113.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 156-57-0
Molecular Formula C2H8ClNS
Molecular Weight (g/mol) 113.60
MDL Number MFCD00012904
InChI Key OGMADIBCHLQMIP-UHFFFAOYSA-N
Synonym cysteamine hydrochloride, 2-aminoethanethiol hydrochloride, cysteamine hcl, cysteaminium chloride, 2-mercaptoethylamine hydrochloride, 2-mercaptoethylamine hcl, bekaptan, mercaptamine hydrochloride, merkamin hydrochloride
PubChem CID 9082
SMILES [H+].[Cl-].NCCS

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 66.0°C to 70.0°C
Color White
Density .75g/mL
Loss on Drying 1% max. (50°C, 4 hrs, 100mm Hg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NCH2CH2SH·HCl
Quantity 500 g
Beilstein 04, 286
Merck Index 15, 2776
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: very soluble in alcohol
IUPAC Name hydrogen 2-aminoethane-1-thiol chloride
Formula Weight 113.61
Percent Purity 98%
Physical Form Crystalline Powder or Pellets
Chemical Name or Material 2-Aminoethanethiol hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-858-1

RUO – Research Use Only