missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminodiphenylmethane 98.0+%, TCI America™
$113.71 - $436.19
Chemical Identifiers
| CAS | 28059-64-5 |
|---|---|
| Molecular Formula | C13H13N |
| Molecular Weight (g/mol) | 183.25 |
| MDL Number | MFCD00007750 |
| InChI Key | DWOBGCPUQNFAFB-UHFFFAOYSA-N |
| Synonym | 2-Benzylaniline |
| PubChem CID | 119805 |
| IUPAC Name | 2-benzylaniline |
| SMILES | NC1=CC=CC=C1CC1=CC=CC=C1 |
Chemical Identifiers
| 28059-64-5 | |
| 183.25 | |
| DWOBGCPUQNFAFB-UHFFFAOYSA-N | |
| 119805 | |
| NC1=CC=CC=C1CC1=CC=CC=C1 |
| C13H13N | |
| MFCD00007750 | |
| 2-Benzylaniline | |
| 2-benzylaniline |
Specifications
| 28059-64-5 | |
| 172°C | |
| MFCD00007750 | |
| 2-Benzylaniline | |
| NC1=CC=CC=C1CC1=CC=CC=C1 | |
| 183.25 | |
| 183.25 | |
| Crystalline Powder |
| 51°C | |
| C13H13N | |
| 5 g | |
| DWOBGCPUQNFAFB-UHFFFAOYSA-N | |
| 2-benzylaniline | |
| 119805 | |
| ≥98.0% (GC,T) | |
| 2-Aminodiphenylmethane |
Safety and Handling
TSCA : No