missing translation for 'onlineSavingsMsg'
Learn More

2-Aminodiphenylmethane 98.0+%, TCI America™

Catalog No. A25195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25195G 5 g
A251925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25195G Supplier TCI America Supplier No. A25195G
Only null left

Chemical Identifiers

CAS 28059-64-5
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.25
MDL Number MFCD00007750
InChI Key DWOBGCPUQNFAFB-UHFFFAOYSA-N
Synonym 2-Benzylaniline
PubChem CID 119805
IUPAC Name 2-benzylaniline
SMILES NC1=CC=CC=C1CC1=CC=CC=C1

Specifications

Melting Point 51°C
Boiling Point 172°C
Quantity 5 g
Formula Weight 183.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminodiphenylmethane