missing translation for 'onlineSavingsMsg'
Learn More

2-Aminocyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. A125825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A125825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A125825G Supplier TCI America Supplier No. A125825G

Chemical Identifiers

CAS 6850-38-0
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD08061325,MFCD08061326,MFCD00191368
InChI Key PQMCFTMVQORYJC-UHFFFAOYNA-N
Synonym 1-Amino-2-hydroxycyclohexane
PubChem CID 23286
IUPAC Name 2-aminocyclohexan-1-ol
SMILES NC1CCCCC1O

Specifications

Quantity 25 g
UN Number 3259
Formula Weight 115.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminocyclohexanol (cis- and trans- mixture)
TSCA No
Recommended Storage Refrigerator