CAS RN 6850-38-0
CAS RN 6850-38-0
IUPAC Name:
2-aminocyclohexan-1-ol
Synonyms:
2-aminocyclohexan-1-ol
Molecular Weight (g/mol):
115.18
Molecular Formula:
C6H13NO
InChi Key:
PQMCFTMVQORYJC-UHFFFAOYNA-N
SMILES:
NC1CCCCC1O
1
–
1
of
1
results
1
2-Aminocyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™
CAS: 6850-38-0 Molecular Formula: C6H13NO Molecular Weight (g/mol): 115.18 MDL Number: MFCD08061325,MFCD08061326,MFCD00191368 InChI Key: PQMCFTMVQORYJC-UHFFFAOYNA-N Synonym: 1-Amino-2-hydroxycyclohexane PubChem CID: 23286 IUPAC Name: 2-aminocyclohexan-1-ol SMILES: NC1CCCCC1O