missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzyl alcohol, 98%, Thermo Scientific™

Catalog No. AC153730100
Change view
Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153730100 10g Glass bottle
AC153730250 25g Glass bottle
AC153731000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC153730100 Supplier Thermo Scientific Chemicals Supplier No. 153730100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 5344-90-1
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD00007749
InChI Key VYFOAVADNIHPTR-UHFFFAOYSA-N
Synonym 2-aminobenzyl alcohol, 2-aminophenyl methanol, 2-aminobenzylalcohol, o-aminobenzyl alcohol, 2-aminobenzenemethanol, benzenemethanol, 2-amino, o-aminobenzylic alcohol, o-hydroxymethyl aniline, benzyl alcohol, o-amino, 2-hydroxymethylaniline
PubChem CID 21439
IUPAC Name (2-aminophenyl)methanol
SMILES C1=CC=C(C(=C1)CO)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Boiling Point 162.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4905
Linear Formula H2NC6H4CH2OH
Quantity 10g
Beilstein 13, 615
Solubility Information Solubility in water: 30g/100g (20°C). Other solubilities: 40g/100g thf (14°C),0.64g/100g toluene (20°C),soluble in chlorinated solvents,soluble in ethanol,ether,very soluble in benzene
Formula Weight 123.15
Percent Purity 98%
Chemical Name or Material 2-Aminobenzyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-293-7

RUO – Research Use Only