missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzoxazole 98.0+%, TCI America™

Catalog No. p-7118065
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27745G 5 g
A277425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27745G Supplier TCI America Supplier No. A27745G
Only null left

Chemical Identifiers

CAS 4570-41-6
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.138
MDL Number MFCD00182850
InChI Key JPBLHOJFMBOCAF-UHFFFAOYSA-N
PubChem CID 20707
IUPAC Name 1,3-benzoxazol-2-amine
SMILES C1=CC=C2C(=C1)N=C(O2)N

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 5 g
Formula Weight 134.14
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminobenzoxazole
RTECSNumber DM4500000
TSCA No