accessibility menu, dialog, popup

UserName

H2NON

CAS RN 4570-41-6

IUPAC Name: 1,3-benzoxazol-2-amine
Synonyms: benzoxazole, 2-amino
Molecular Weight (g/mol): 134.14
Molecular Formula: C7H6N2O
InChi Key: JPBLHOJFMBOCAF-UHFFFAOYSA-N
SMILES: NC1=NC2=CC=CC=C2O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results