missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminobenzothiazole 98.0+%, TCI America™
$142.46 - $531.34
Chemical Identifiers
| CAS | 136-95-8 |
|---|---|
| Molecular Formula | C7H6N2S |
| Molecular Weight (g/mol) | 150.199 |
| MDL Number | MFCD00005785 |
| InChI Key | UHGULLIUJBCTEF-UHFFFAOYSA-N |
| Synonym | 2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine |
| PubChem CID | 8706 |
| IUPAC Name | 1,3-benzothiazol-2-amine |
| SMILES | C1=CC=C2C(=C1)N=C(S2)N |
Chemical Identifiers
| 136-95-8 | |
| 150.199 | |
| UHGULLIUJBCTEF-UHFFFAOYSA-N | |
| 8706 | |
| C1=CC=C2C(=C1)N=C(S2)N |
| C7H6N2S | |
| MFCD00005785 | |
| 2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine | |
| 1,3-benzothiazol-2-amine |
Specifications
| 136-95-8 | |
| White-Yellow | |
| MFCD00005785 | |
| 2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine | |
| C1=CC=C2C(=C1)N=C(S2)N | |
| 150.199 | |
| 150.20 | |
| Crystalline Powder |
| 130°C | |
| C7H6N2S | |
| 25 g | |
| UHGULLIUJBCTEF-UHFFFAOYSA-N | |
| 1,3-benzothiazol-2-amine | |
| 8706 | |
| ≥98.0% (HPLC,T) | |
| 2-Aminobenzothiazole |
Safety and Handling
EINECSNumber : (5)-0234
RTECSNumber : DL1050000
TSCA : Yes