missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzothiazole 98.0+%, TCI America™

Catalog No. A027725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A027725G 25 g
A0277100G 100 g
A0277500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A027725G Supplier TCI America Supplier No. A027725G

Chemical Identifiers

CAS 136-95-8
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.199
MDL Number MFCD00005785
InChI Key UHGULLIUJBCTEF-UHFFFAOYSA-N
Synonym 2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine
PubChem CID 8706
IUPAC Name 1,3-benzothiazol-2-amine
SMILES C1=CC=C2C(=C1)N=C(S2)N

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 25 g
Formula Weight 150.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminobenzothiazole
EINECSNumber (5)-0234
RTECSNumber DL1050000
TSCA Yes