missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzothiazole, 98%

Catalog No. AAA1352122
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1352122 100 g
AAA1352136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1352122 Supplier Thermo Scientific Chemicals Supplier No. A1352122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 136-95-8 | C7H6N2S | 150.199 g/mol

2-Aminobenzothiazole was used in the synthesis of cobalt(II) picrate mixed-ligand complexes. It was used to study adsorption of biologically significant 2-aminobenzothiazole molecules on colloidal silver particles using surface-enhanced raman scattering spectroscopy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzothiazole was used in the synthesis of cobalt(II) picrate mixed-ligand complexes. It was used to study adsorption of biologically significant 2-aminobenzothiazole molecules on colloidal silver particles using surface-enhanced raman scattering spectroscopy

Solubility
Soluble in alcohol, chloroform, diethyl ether.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 136-95-8
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.199
MDL Number MFCD00005785
InChI Key UHGULLIUJBCTEF-UHFFFAOYSA-N
Synonym 2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine
PubChem CID 8706
IUPAC Name 1,3-benzothiazol-2-amine
SMILES C1=CC=C2C(=C1)N=C(S2)N

Specifications

Melting Point 126°C to 129°C
Quantity 100 g
Beilstein 116315
Merck Index 14,424
Solubility Information Soluble in alcohol,chloroform,diethyl ether.
Formula Weight 150.2
Percent Purity 98%
Chemical Name or Material 2-Aminobenzothiazole
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 205-268-4
RTECSNumber DL1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only