missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzothiazole, 97%

CAS: 136-95-8 | C7H6N2S | 150.2 g/mol

$88.90 - $88.90

Chemical Identifiers

CAS 136-95-8
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.2
MDL Number MFCD00005785
InChI Key UHGULLIUJBCTEF-UHFFFAOYSA-N
Synonym 2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine
PubChem CID 8706
IUPAC Name 1,3-benzothiazol-2-amine
SMILES C1=CC=C2C(=C1)N=C(S2)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC148811000
SDS View Documents
Thermo Scientific Chemicals
148811000
100 g Plastic bottle
Each for $88.90
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

136-95-8
150.2
UHGULLIUJBCTEF-UHFFFAOYSA-N
8706
C1=CC=C2C(=C1)N=C(S2)N
C7H6N2S
MFCD00005785
2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine
1,3-benzothiazol-2-amine

Specifications

136-95-8
100.0
Beige to Gray
97%
C7H6N2S
100 g
15, 419
Solubility in water: sparingly soluble in water. Other solubilities: soluble in concentrated acids,freely soluble in alcohol,ether and chloroform
C1=CC=C2C(=C1)N=C(S2)N
150.2
150.2
Powder or Flakes
96.0
129°C to 134°C
Authentic
Plastic bottle
MFCD00005785
27, 182
2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine
UHGULLIUJBCTEF-UHFFFAOYSA-N
1,3-benzothiazol-2-amine
8706
97%
2-Aminobenzothiazole, 97%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-268-4

RTECSNumber : DL1050000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only