missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzophenone, 98%

CAS: 2835-77-0 | C13H11NO | 197.24 g/mol

$596.11 - $596.11

Chemical Identifiers

CAS 2835-77-0
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00007713
InChI Key MAOBFOXLCJIFLV-UHFFFAOYSA-N
Synonym 2-aminobenzophenone, 2-benzoylaniline, o-benzoylaniline, o-aminobenzophenone, 2-aminophenyl phenyl methanone, methanone, 2-aminophenyl phenyl, benzophenone, 2-amino, 2-amino-phenyl-phenyl-methanone, 2-aminophenyl phenyl ketone, methanone, aminophenyl phenyl
PubChem CID 76080
IUPAC Name (2-aminophenyl)-phenylmethanone
SMILES NC1=CC=CC=C1C(=O)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC103161000
SDS View Documents
Thermo Scientific Chemicals
103161000
100 g Glass bottle
Each for $596.11
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2835-77-0
197.24
MAOBFOXLCJIFLV-UHFFFAOYSA-N
76080
NC1=CC=CC=C1C(=O)C1=CC=CC=C1
C13H11NO
MFCD00007713
2-aminobenzophenone, 2-benzoylaniline, o-benzoylaniline, o-aminobenzophenone, 2-aminophenyl phenyl methanone, methanone, 2-aminophenyl phenyl, benzophenone, 2-amino, 2-amino-phenyl-phenyl-methanone, 2-aminophenyl phenyl ketone, methanone, aminophenyl phenyl
(2-aminophenyl)-phenylmethanone

Specifications

2835-77-0
100.0
Golden-Yellow
Authentic
Glass bottle
H2NC6H4COC6H5
100 g
2-aminobenzophenone, 2-benzoylaniline, o-benzoylaniline, o-aminobenzophenone, 2-aminophenyl phenyl methanone, methanone, 2-aminophenyl phenyl, benzophenone, 2-amino, 2-amino-phenyl-phenyl-methanone, 2-aminophenyl phenyl ketone, methanone, aminophenyl phenyl
MAOBFOXLCJIFLV-UHFFFAOYSA-N
(2-aminophenyl)-phenylmethanone
76080
98%
2-Aminobenzophenone, 98%
97.5
105.0°C to 108.0°C
240°C
98%
C13H11NO
MFCD00007713
14, 76
Solubility in water: practically insoluble. Other solubilities: 40g/L methanol (20°C),soluble in chloroform
NC1=CC=CC=C1C(=O)C1=CC=CC=C1
197.24
197.24
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 220-613-9

SDS
Documents

Documents

RUO – Research Use Only