missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzophenone, 98%

CAS: 2835-77-0 | C13H11NO | 197.24 g/mol

Supplier:  Thermo Scientific Chemicals A1258018

 View more versions of this product

Catalog No. AAA1258018


Only null left
Explore available promotions

Description

Description

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

Solubility
Insoluble in water. Soluble in DMSO, Methanol.

Notes
Store at room temperature. Ensure good ventilation.
Specifications

Specifications

2-Aminobenzophenone
105°C to 108°C
240°C (464°F)
MFCD00007713
743425
Insoluble in water. Soluble in DMSO,Methanol.
NC1=CC=CC=C1C(=O)C1=CC=CC=C1
197.24
197.24
2835-77-0
280°C
C13H11NO
50 g
2-aminobenzophenone, 2-benzoylaniline, o-benzoylaniline, o-aminobenzophenone, 2-aminophenyl phenyl methanone, methanone, 2-aminophenyl phenyl, benzophenone, 2-amino, 2-amino-phenyl-phenyl-methanone, 2-aminophenyl phenyl ketone, methanone, aminophenyl phenyl
MAOBFOXLCJIFLV-UHFFFAOYSA-N
(2-aminophenyl)-phenylmethanone
76080
98%

Chemical Identifiers

2835-77-0
197.24
MAOBFOXLCJIFLV-UHFFFAOYSA-N
76080
NC1=CC=CC=C1C(=O)C1=CC=CC=C1
C13H11NO
MFCD00007713
2-aminobenzophenone, 2-benzoylaniline, o-benzoylaniline, o-aminobenzophenone, 2-aminophenyl phenyl methanone, methanone, 2-aminophenyl phenyl, benzophenone, 2-amino, 2-amino-phenyl-phenyl-methanone, 2-aminophenyl phenyl ketone, methanone, aminophenyl phenyl
(2-aminophenyl)-phenylmethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 220-613-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only