missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzophenone, 98%

Catalog No. AC103160250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103160250 25 g Glass bottle
AC103161000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC103160250 Supplier Thermo Scientific Chemicals Supplier No. 103160250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2835-77-0 | C13H11NO | 197.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2835-77-0
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00007713
InChI Key MAOBFOXLCJIFLV-UHFFFAOYSA-N
Synonym 2-aminobenzophenone, 2-benzoylaniline, o-benzoylaniline, o-aminobenzophenone, 2-aminophenyl phenyl methanone, methanone, 2-aminophenyl phenyl, benzophenone, 2-amino, 2-amino-phenyl-phenyl-methanone, 2-aminophenyl phenyl ketone, methanone, aminophenyl phenyl
PubChem CID 76080
IUPAC Name (2-aminophenyl)-phenylmethanone
SMILES NC1=CC=CC=C1C(=O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 105.0°C to 108.0°C
Color Golden-Yellow
Flash Point 240°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NC6H4COC6H5
Quantity 25 g
Beilstein 14, 76
Solubility Information Solubility in water: practically insoluble. Other solubilities: 40g/L methanol (20°C),soluble in chloroform
Formula Weight 197.24
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Aminobenzophenone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 220-613-9

RUO – Research Use Only