missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzenethiol 97.0+%, TCI America™

Catalog No. A0267100ML
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
A0267100ML 100 mL
A026725ML 25 mL
A0267500ML 500 mL
3 options

Catalog No. A0267100ML

Supplier: TCI America A0267100ML

Only null left

Chemical Identifiers

CAS 137-07-5
Molecular Formula C6H7NS
Molecular Weight (g/mol) 125.19
MDL Number MFCD00007702
InChI Key VRVRGVPWCUEOGV-UHFFFAOYSA-N
Synonym 2-aminothiophenol, o-aminothiophenol, 2-mercaptoaniline, o-mercaptoaniline, o-aminobenzenethiol, 2-mercaptaniline, benzenethiol, 2-amino, 2-amino thiophenol, 1-amino-2-mercaptobenzene, 2-aminobenzene-1-thiol
PubChem CID 8713
IUPAC Name 2-aminobenzene-1-thiol
SMILES NC1=CC=CC=C1S

Specifications

Color Yellow
Boiling Point 234°C
Quantity 100 mL
UN Number 3267
Formula Weight 125.19
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Aminobenzenethiol
EINECSNumber (3)-2899
RTECSNumber DC0600000
TSCA Yes