missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzeneboronic acid, 96%

Catalog No. p-7047219
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1806903 1 g
AAL1806906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1806903 Supplier Thermo Scientific Chemicals Supplier No. L1806903
Only null left

CAS: 5570-18-3 | C6H8BNO2 | 136.945 g/mol

2-Aminobenzeneboronic acid derivative is used in the synthesis of quinolines by reacting with alpha, beta unsaturated ketones. Its hydrochloride derivative is used in the preparation of designed boronate ligands for glucose-selective holographic sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzeneboronic acid derivative is used in the synthesis of quinolines by reacting with alpha, beta unsaturated ketones. Its hydrochloride derivative is used in the preparation of designed boronate ligands for glucose-selective holographic sensors.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 5570-18-3
Molecular Formula C6H8BNO2
Molecular Weight (g/mol) 136.945
MDL Number MFCD01074645
InChI Key DIRRKLFMHQUJCM-UHFFFAOYSA-N
Synonym 2-aminophenyl boronic acid, 2-aminobenzeneboronic acid, o-aminophenylboronic acid, 2-aminophenylboronicacid, 2-boronoaniline, pubchem9885, acmc-1apjj, 2-amino-phenylboronic acid, 2-amino benzeneboronic acid
PubChem CID 2773216
IUPAC Name (2-aminophenyl)boronic acid
SMILES B(C1=CC=CC=C1N)(O)O

Specifications

Melting Point 188°C to 190°C
Quantity 1 g
Beilstein 2962939
Formula Weight 136.95
Percent Purity 96%
Chemical Name or Material 2-Aminobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only