accessibility menu, dialog, popup

UserName

CAS RN 5570-18-3

IUPAC Name: N'-[(Z)-(2,5-dimethoxyphenyl)methylidene]-2-[(4-methylphenyl)amino]acetohydrazide
Synonyms: N'-[(Z)-(2,5-dimethoxyphenyl)methylidene]-2-[(4-methylphenyl)amino]acetohydrazide
Molecular Weight (g/mol): 327.38
Molecular Formula: C18H21N3O3
InChi Key: PTSWLQVBIWPGOF-JAIQZWGSSA-N
SMILES: COC1=CC=C(OC)C(\C=N/NC(=O)CNC2=CC=C(C)C=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results