missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzamidoxime, 97%

Catalog No. p-7034125
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5178203 1 g
AAH5178206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5178203 Supplier Thermo Scientific Chemicals Supplier No. H5178203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16348-49-5 | C7H9N3O | 151.169 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16348-49-5
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.169
MDL Number MFCD00492723
InChI Key CFZHYRNQLHEHJS-UHFFFAOYSA-N
Synonym 2-aminobenzamide oxime, 2-aminobenzamidoxime, 2-amino-n'-hydroxybenzimidamide, 2-amino-n-hydroxybenzimidamide, z-2-amino-n'-hydroxybenzimidamide, e-2-amino-n'-hydroxybenzenecarboximidamide
PubChem CID 9628840
IUPAC Name 2-amino-N'-hydroxybenzenecarboximidamide
SMILES C1=CC=C(C(=C1)C(=NO)N)N

Specifications

Melting Point 71°C to 75°C
Quantity 1 g
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material 2-Aminobenzamidoxime
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only