missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzamide 98.0+%, TCI America™

Catalog No. A026225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A026225G 25 g
A0262100G 100 g
A0262500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A026225G Supplier TCI America Supplier No. A026225G

Chemical Identifiers

CAS 88-68-6
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00007981
InChI Key PXBFMLJZNCDSMP-UHFFFAOYSA-N
Synonym anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van
PubChem CID 6942
IUPAC Name 2-aminobenzamide
SMILES C1=CC=C(C(=C1)C(=O)N)N

Specifications

Melting Point 111°C
Color White
Quantity 25 g
Formula Weight 136.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminobenzamide
EINECSNumber (3)-4561
RTECSNumber CU8993000
TSCA Yes