missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoazotoluene 97.0+%, TCI America™

Supplier:  TCI America T026125G

 View more versions of this product

Catalog No. T026125G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Aminoazotoluene
102°C
MFCD00007733
o-aminoazotoluene, fast garnet gbc base, solvent yellow 3, c.i. solvent yellow 3, toluazotoluidine, o-aminoazotoluol, fast oil yellow, somalia yellow r, fast yellow at, fat yellow b
CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C
225.295
225.30
Crystalline Powder
97-56-3
C14H15N3
25 g
PFRYFZZSECNQOL-UHFFFAOYSA-N
2-methyl-4-[(2-methylphenyl)diazenyl]aniline
7340
≥97.0% (HPLC,T)

Chemical Identifiers

97-56-3
225.295
PFRYFZZSECNQOL-UHFFFAOYSA-N
7340
CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C
C14H15N3
MFCD00007733
o-aminoazotoluene, fast garnet gbc base, solvent yellow 3, c.i. solvent yellow 3, toluazotoluidine, o-aminoazotoluol, fast oil yellow, somalia yellow r, fast yellow at, fat yellow b
2-methyl-4-[(2-methylphenyl)diazenyl]aniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-2299&(5)-5002

RTECSNumber : XU8800000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions