missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminoazotoluene 97.0+%, TCI America™
$34.66 - $34.66
Chemical Identifiers
| CAS | 97-56-3 |
|---|---|
| Molecular Formula | C14H15N3 |
| Molecular Weight (g/mol) | 225.295 |
| MDL Number | MFCD00007733 |
| InChI Key | PFRYFZZSECNQOL-UHFFFAOYSA-N |
| Synonym | o-aminoazotoluene, fast garnet gbc base, solvent yellow 3, c.i. solvent yellow 3, toluazotoluidine, o-aminoazotoluol, fast oil yellow, somalia yellow r, fast yellow at, fat yellow b |
| PubChem CID | 7340 |
| IUPAC Name | 2-methyl-4-[(2-methylphenyl)diazenyl]aniline |
| SMILES | CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C |
Chemical Identifiers
| 97-56-3 | |
| 225.295 | |
| PFRYFZZSECNQOL-UHFFFAOYSA-N | |
| 7340 | |
| CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C |
| C14H15N3 | |
| MFCD00007733 | |
| o-aminoazotoluene, fast garnet gbc base, solvent yellow 3, c.i. solvent yellow 3, toluazotoluidine, o-aminoazotoluol, fast oil yellow, somalia yellow r, fast yellow at, fat yellow b | |
| 2-methyl-4-[(2-methylphenyl)diazenyl]aniline |
Specifications
| 97-56-3 | |
| C14H15N3 | |
| 25 g | |
| PFRYFZZSECNQOL-UHFFFAOYSA-N | |
| 2-methyl-4-[(2-methylphenyl)diazenyl]aniline | |
| 7340 | |
| ≥97.0% (HPLC,T) | |
| 2-Aminoazotoluene |
| 102°C | |
| MFCD00007733 | |
| o-aminoazotoluene, fast garnet gbc base, solvent yellow 3, c.i. solvent yellow 3, toluazotoluidine, o-aminoazotoluol, fast oil yellow, somalia yellow r, fast yellow at, fat yellow b | |
| CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C | |
| 225.295 | |
| 225.30 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-2299&(5)-5002
RTECSNumber : XU8800000
TSCA : Yes