missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminoadenosine 98.0+%, TCI America™
$80.16 - $395.17
Chemical Identifiers
| CAS | 2096-10-8 |
|---|---|
| Molecular Formula | C10H14N6O4 |
| Molecular Weight (g/mol) | 282.26 |
| MDL Number | MFCD00053556 |
| InChI Key | ZDTFMPXQUSBYRL-SBTUYZBDNA-N |
| Synonym | 2-aminoadenosine, 2-amine adenosine, adenosine, 2-amino, 2,6-diaminopurine riboside, 2,6-diaminonebularine, 2,6-diaminopurinosine, 2r,3r,4s,5r-2-2,6-diaminopurin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 2,6-diamino-9-beta-d-ribofuranosyl purine, pubchem14156, ksc202m1p |
| PubChem CID | 72200 |
| ChEBI | CHEBI:1014 |
| IUPAC Name | (2R,3R,4S,5R)-2-(2,6-diamino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
| SMILES | NC1=NC(N)=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=N1 |
Chemical Identifiers
| 2096-10-8 | |
| 282.26 | |
| ZDTFMPXQUSBYRL-SBTUYZBDNA-N | |
| 72200 | |
| (2R,3R,4S,5R)-2-(2,6-diamino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
| C10H14N6O4 | |
| MFCD00053556 | |
| 2-aminoadenosine, 2-amine adenosine, adenosine, 2-amino, 2,6-diaminopurine riboside, 2,6-diaminonebularine, 2,6-diaminopurinosine, 2r,3r,4s,5r-2-2,6-diaminopurin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 2,6-diamino-9-beta-d-ribofuranosyl purine, pubchem14156, ksc202m1p | |
| CHEBI:1014 | |
| NC1=NC(N)=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=N1 |
Specifications
| 2096-10-8 | |
| White-Yellow | |
| MFCD00053556 | |
| 2-aminoadenosine, 2-amine adenosine, adenosine, 2-amino, 2,6-diaminopurine riboside, 2,6-diaminonebularine, 2,6-diaminopurinosine, 2r,3r,4s,5r-2-2,6-diaminopurin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 2,6-diamino-9-beta-d-ribofuranosyl purine, pubchem14156, ksc202m1p | |
| NC1=NC(N)=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=N1 | |
| 282.26 | |
| CHEBI:1014 | |
| ≥98.0% (HPLC,T) | |
| 2-Aminoadenosine |
| 252°C | |
| C10H14N6O4 | |
| 5 g | |
| ZDTFMPXQUSBYRL-SBTUYZBDNA-N | |
| (2R,3R,4S,5R)-2-(2,6-diamino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol | |
| 72200 | |
| 282.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : No