missing translation for 'onlineSavingsMsg'
Learn More

2'-Aminoacetanilide 98.0+%, TCI America™

Catalog No. p-7117899
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24265G 5 g
A242625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24265G Supplier TCI America Supplier No. A24265G
Only null left

Chemical Identifiers

CAS 34801-09-7
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD00210388
InChI Key MPXAYYWSDIKNTP-UHFFFAOYSA-N
Synonym n-2-aminophenyl acetamide, 2'-aminoacetanilide, o-aminoacetanilide, acetamide, n-2-aminophenyl, unii-lb34xrq95v, lb34xrq95v, acetamide, n-2-aminophenyl-9ci, n-acetyl-2-aminoaniline, n-acetyl-o-phenylenediamine, 2-aminoactanilide
PubChem CID 11149
IUPAC Name N-(2-aminophenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1N

Specifications

Melting Point 134°C
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Aminoacetanilide
EINECSNumber (3)-0244
TSCA No