missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-p-cresol 98.0+%, TCI America™

Catalog No. A033025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A033025G 25 g
A0330100G 100 g
A0330500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A033025G Supplier TCI America Supplier No. A033025G

Chemical Identifiers

CAS 95-84-1
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00007699
InChI Key ZMXYNJXDULEQCK-UHFFFAOYSA-N
Synonym 2-amino-p-cresol, 2-hydroxy-5-methylaniline, phenol, 2-amino-4-methyl, 3-amino-4-hydroxytoluene, 6-hydroxy-m-toluidine, 4-methyl-2-aminophenol, 5-methyl-2-hydroxyaniline, 2-amino-4-cresol, p-cresol, 2-amino, 2-amino-4-methyl-phenol
PubChem CID 7264
IUPAC Name 2-amino-4-methylphenol
SMILES CC1=CC=C(O)C(N)=C1

Specifications

Melting Point 135°C
Quantity 25 g
Formula Weight 123.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-p-cresol
EINECSNumber (3)-0707&(3)-0676
RTECSNumber SJ6078000
TSCA Yes