missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide 98.0+%, TCI America™

Catalog No. p-7118064
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27731G 1 g
A27735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27731G Supplier TCI America Supplier No. A27731G

Chemical Identifiers

CAS 302964-24-5
Molecular Formula C11H10ClN3OS
Molecular Weight (g/mol) 267.731
MDL Number MFCD10000630
InChI Key VVOXTERFTAJMAA-UHFFFAOYSA-N
PubChem CID 21911644
IUPAC Name 2-amino-N-(2-chloro-6-methylphenyl)-1,3-thiazole-5-carboxamide
SMILES CC1=C(C(=CC=C1)Cl)NC(=O)C2=CN=C(S2)N

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 1 g
Formula Weight 267.73
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide