missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-9-fluorenone 98.0+%, TCI America™

Catalog No. A10401G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10401G 1 g
A10405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10401G Supplier TCI America Supplier No. A10401G

Chemical Identifiers

CAS 3096-57-9
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.22
MDL Number MFCD00001157
InChI Key SJODITPGMMSNRF-UHFFFAOYSA-N
PubChem CID 18357
IUPAC Name 2-amino-9H-fluoren-9-one
SMILES NC1=CC=C2C3=CC=CC=C3C(=O)C2=C1

Specifications

Melting Point 156°C
Color Purple-Red
Quantity 1 g
Synonym 2-amino-9-fluorenone, 2-amino-9h-fluoren-9-one, 2-aminofluorenone, 9h-fluoren-9-one, 2-amino, 9h-fluoren-9-one,2-amino, fluoren-9-one, 2-amino, 9h-fluoren-9-one, 2-amino-9ci, 2-amino-fluorenon, 2-aminofluorenone-9, 2-aminofluorene-9-one
Formula Weight 195.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-9-fluorenone
RTECSNumber LL8980200
TSCA No