missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-9-fluorenone, 98%

Catalog No. p-7036711
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5504803 1 g
AAH5504806 5 g
AAH5504814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5504803 Supplier Thermo Scientific Chemicals Supplier No. H5504803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3096-57-9 | C13H9NO | 195.22 g/mol

2-Amino-9-fluorenone is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-9-fluorenone is used as an active pharmaceutical intermediate.

Solubility
Soluble in alcohol and ether. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3096-57-9
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.22
MDL Number MFCD00001157
InChI Key SJODITPGMMSNRF-UHFFFAOYSA-N
Synonym 2-amino-9-fluorenone, 2-amino-9h-fluoren-9-one, 2-aminofluorenone, 9h-fluoren-9-one, 2-amino, 9h-fluoren-9-one,2-amino, fluoren-9-one, 2-amino, 9h-fluoren-9-one, 2-amino-9ci, 2-amino-fluorenon, 2-aminofluorenone-9, 2-aminofluorene-9-one
PubChem CID 18357
IUPAC Name 2-aminofluoren-9-one
SMILES NC1=CC=C2C3=CC=CC=C3C(=O)C2=C1

Specifications

Melting Point 157°C to 160°C
Quantity 1 g
Solubility Information Soluble in alcohol and ether. Insoluble in water.
Formula Weight 195.22
Percent Purity 98%
Chemical Name or Material 2-Amino-9-fluorenone
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 221-445-9
RTECSNumber LL8980200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only