missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-9,9-dimethylfluorene 99.0+%, TCI America™

Catalog No. A2634200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2634200MG 200 mg
A26341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2634200MG Supplier TCI America Supplier No. A2634200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108714-73-4
Molecular Formula C15H15N
Molecular Weight (g/mol) 209.292
MDL Number MFCD09953790
InChI Key GUTJITRKAMCHSD-UHFFFAOYSA-N
Synonym 2-amino-9,9-dimethylfluorene, 9,9-dimethyl-9h-fluoren-2-amine, 2-amino-9,9-dimethyl-9h-fluorene, 9,9-dimethyl-9h-fluoren-2-ylamine, 9,9-dimethyl9h-fluorene-2-amine, pubchem23234, ksc495c6d, 9,9-dimethyl-2-aminofluorene, 2-amino-9,9-dimethyl fluorene, 9,9-dimethyl-fluoren-2-amine
PubChem CID 22617093
IUPAC Name 9,9-dimethylfluoren-2-amine
SMILES CC1(C2=CC=CC=C2C3=C1C=C(C=C3)N)C

Specifications

Melting Point 168°C
Quantity 200 mg
Formula Weight 209.29
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-9,9-dimethylfluorene