missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-9,9-dimethylfluorene 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 168°C |
| Quantity | 1 g |
| Formula Weight | 209.29 |
| Percent Purity | ≥99.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Amino-9,9-dimethylfluorene |
Safety and Handling
| TSCA | No |