missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals

Catalog No. p-7056033
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6477106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6477106 Supplier Thermo Scientific Chemicals Supplier No. H6477106

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108714-73-4
Molecular Formula C15H15N
Molecular Weight (g/mol) 209.292
MDL Number MFCD09953790
InChI Key GUTJITRKAMCHSD-UHFFFAOYSA-N
Synonym 2-amino-9,9-dimethylfluorene, 9,9-dimethyl-9h-fluoren-2-amine, 2-amino-9,9-dimethyl-9h-fluorene, 9,9-dimethyl-9h-fluoren-2-ylamine, 9,9-dimethyl9h-fluorene-2-amine, pubchem23234, ksc495c6d, 9,9-dimethyl-2-aminofluorene, 2-amino-9,9-dimethyl fluorene, 9,9-dimethyl-fluoren-2-amine
PubChem CID 22617093
IUPAC Name 9,9-dimethylfluoren-2-amine
SMILES CC1(C2=CC=CC=C2C3=C1C=C(C=C3)N)C

Specifications

Color White
Quantity 5 g
Beilstein 15506865
Formula Weight 209.29
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-9,9-dimethylfluorene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only