missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-7-bromofluorene, tech. 90%

Catalog No. p-7036907
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5537003 1 g
AAH5537006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5537003 Supplier Thermo Scientific Chemicals Supplier No. H5537003

CAS: 6638-60-4 | C13H10BrN | 260.134 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Sparingly soluble in water. (0.724 mg/L)

Notes
Incompatible with oxidizing agents. Store at room temperature.

Chemical Identifiers

CAS 6638-60-4
Molecular Formula C13H10BrN
Molecular Weight (g/mol) 260.134
MDL Number MFCD00001127
InChI Key RJWYTISHBMNMOZ-UHFFFAOYSA-N
Synonym 2-amino-7-bromofluorene, 2-bromo-7-aminofluorene, fluoren-2-amine, 7-bromo, 7-bromo-9h-fluoren-2-ylamine, 9h-fluoren-2-amine, 7-bromo, 7-bromofluorene-2-ylamine, acmc-20a5iw, 2-amino-7-bromo-9h-fluorene, 9h-fluoren-2-amine,7-bromo, 7-bromanyl-9h-fluoren-2-amine
PubChem CID 23126
IUPAC Name 7-bromo-9H-fluoren-2-amine
SMILES C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)Br

Specifications

Melting Point 148°C to 149°C
Quantity 1 g
Solubility Information Sparingly soluble in water. (0.724mg/L)
Formula Weight 260.13
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Amino-7-bromofluorene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber LL5150050
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only